Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029492.4 | NP_083768.4 | 1 | 159 | ||
GenBank mRNA | AK017158.1 | - | 1 | 160 | ||
AK020965.1 | - | 1 | 35 | |||
AK031184.1 | - | 1 | 118 | |||
AK049270.1 | - | 1 | 139 | |||
AK139216.1 | - | 1 | 118 | |||
AK140182.1 | - | 1 | 120 | |||
AK151840.1 | - | 1 | 174 | |||
AK163174.1 | - | 1 | 157 | |||
AK170017.1 | - | 1 | 177 | |||
AK170139.1 | - | 1 | 100 | |||
AY668951.1 | - | 1 | 77 | |||
BC019536.1 | - | 1 | 176 | 4039734 |