Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agmat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081408.1 | NP_001074877.1 | 2 | 528 | ||
GenBank mRNA | AK017155.1 | - | 2 | 427 | ||
BC115693.2 | - | 2 | 425 | 40104399 |