Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172672.2 | NP_766260.2 | 14 | 2713 | ||
GenBank mRNA | AK076333.1 | - | 13 | 1657 | ||
AK220303.1 | - | 14 | 2733 | |||
BC079548.1 | - | 14 | 2175 |