Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029519.3 | NP_083795.2 | 2 | 637 | ||
GenBank mRNA | AK075712.1 | - | 2 | 597 | ||
AK140318.1 | - | 2 | 649 | |||
BC043066.1 | - | 2 | 608 | 5709337 | ||
BC049084.1 | - | 2 | 637 | 5706690 | ||
BC053003.1 | - | 2 | 637 | 5706690 | ||
BC067194.1 | - | 2 | 653 |