Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080420.2 | NP_536345.2 | 13 | 2450 | ||
GenBank mRNA | AK018070.1 | - | 5 | 933 | ||
BC016212.1 | - | 13 | 2457 | 4924899 |