Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080420.2 | NP_536345.2 | 9, 10 | 1634 | ||
GenBank mRNA | AK018070.1 | - | 1, 2 | 118 | ||
AK162992.1 | - | 1, 2 | 82 | |||
BC016212.1 | - | 9, 10 | 1641 | 4924899 |