Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081059.3 | NP_001074528.1 | 6 | 812 | ||
GenBank mRNA | AK018089.1 | - | 6 | 783 | ||
AK041849.1 | - | 6 | 813 | |||
AK217814.1 | - | 3 | 127 | |||
BC113195.1 | - | 7 | 550 |