Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172424.4 | NP_766012.3 | 24 | 5462 | ||
GenBank mRNA | AK122419.1 | - | 15 | 3750 | ||
AK154895.1 | - | 24 | 5442 | |||
AK170157.1 | - | 3 | 324 | |||
AY338464.1 | - | 24 | 5439 | |||
BC040283.1 | - | 4 | 1953 | |||
BC049985.1 | - | 2 | 688 | |||
BC052320.1 | - | 8 | 2149 | |||
BC072653.1 | - | 17 | 4385 |