Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029553.3 | NP_083829.1 | 10, 11 | 1126 | ||
NM_198311.1 | NP_938053.1 | 11, 12 | 1118 | |||
GenBank mRNA | AK002597.1 | - | 10, 11 | 1112 | ||
AK032748.1 | - | 10, 11 | 1088 | |||
AK081697.1 | - | 10, 11 | 1093 | |||
BC017523.1 | - | 11, 12 | 1118 | 4527657 |