Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029580.3 | NP_083856.1 | 2 | 493 | ||
GenBank mRNA | AK011576.1 | - | 2 | 494 | ||
AK049127.1 | - | 2 | 492 | |||
AK075622.1 | - | 1 | 288 | |||
AK220389.1 | - | 1 | 240 | |||
BC012521.1 | - | 2 | 494 | 2648293 |