Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtif3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256100.1 | NP_001243029.1 | 3 | 383 | ||
NM_001256101.1 | NP_001243030.1 | 3 | 372 | |||
NM_001256102.1 | NP_001243031.1 | 2 | 212 | |||
NM_029581.4 | NP_083857.1 | 2 | 223 | |||
GenBank mRNA | AK012733.1 | - | 3 | 350 | ||
AK147090.1 | - | 2 | 201 | |||
BC052439.1 | - | 2 | 177 | 6406303 | ||
BC115684.1 | - | 2 | 164 | 40104230 | ||
BC115685.1 | - | 3 | 335 | 40104232 | ||
BY284518.1 | - | 2 | 212 |