Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029614.3 | NP_083890.2 | 2 | 1126 | ||
GenBank mRNA | AK009847.1 | - | 2 | 1085 | ||
AK033671.1 | - | 2 | 1102 | |||
AK034439.1 | - | 2 | 1092 | |||
AK078518.1 | - | 2 | 1095 | |||
AK156348.1 | - | 2 | 1093 | |||
BC018517.1 | - | 2 | 1144 | 3597686 |