Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110002H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029623.2 | NP_083899.1 | 18 | 1721 | ||
GenBank mRNA | AF003352.1 | - | 6 | 607 | ||
AK013979.1 | - | 12 | 1107 | |||
AK144623.1 | - | 18 | 1722 | |||
AK154240.1 | - | 18 | 1716 | |||
AK159691.1 | - | 18 | 1716 | |||
BC012285.1 | - | 17 | 1884 | |||
BC021846.1 | - | 18 | 1704 | 5027035 | ||
BC044861.1 | - | 3 | 3362 |