Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081960.1 | NP_001075429.1 | 24, 25 | 2490 | ||
NM_001114347.1 | NP_001107819.1 | 30, 31 | 3232 | |||
NM_001286600.1 | NP_001273529.1 | 25, 26 | 2541 | |||
NM_001286601.1 | NP_001273530.1 | 25, 26 | 2507 | |||
NM_001286602.1 | NP_001273531.1 | 25, 26 | 2553 | |||
NM_029633.2 | NP_083909.2 | 23, 24 | 2487 | |||
GenBank mRNA | AB093252.1 | - | 21, 22 | 2076 | ||
AK043551.1 | - | 23, 24 | 2487 | |||
BC117963.1 | - | 24, 25 | 2478 | 40098296 | ||
BC117964.1 | - | 25, 26 | 2529 | 40098298 | ||
BC131791.1 | - | 24, 25 | 2478 | 40098299 | ||
BC131792.1 | - | 25, 26 | 2495 | 40098301 | ||
BC141415.1 | - | 25, 26 | 2539 | 9056028 |