Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081960.1 | NP_001075429.1 | 32, 33 | 3805 | ||
NM_001114347.1 | NP_001107819.1 | 38, 39 | 4547 | |||
NM_001286600.1 | NP_001273529.1 | 33, 34 | 3856 | |||
NM_001286601.1 | NP_001273530.1 | 33, 34 | 3822 | |||
NM_001286602.1 | NP_001273531.1 | 33, 34 | 3868 | |||
NM_029633.2 | NP_083909.2 | 31, 32 | 3802 | |||
GenBank mRNA | AB093252.1 | - | 29, 30 | 3391 | ||
AJ276961.1 | - | 14, 15 | 1805 | |||
AK043551.1 | - | 31, 32 | 3802 | |||
BC026688.1 | - | 3, 4 | 433 | |||
BC117963.1 | - | 32, 33 | 3793 | 40098296 | ||
BC117964.1 | - | 33, 34 | 3844 | 40098298 | ||
BC131791.1 | - | 32, 33 | 3793 | 40098299 | ||
BC131792.1 | - | 33, 34 | 3810 | 40098301 | ||
BC141415.1 | - | 33, 34 | 3854 | 9056028 |