Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033175.2 | NP_001028347.1 | 4 | 550 | ||
GenBank mRNA | AK154414.1 | - | 4 | 551 | ||
AK154551.1 | - | 4 | 542 | |||
BC116794.1 | - | 3 | 376 | |||
BC116798.1 | - | 3 | 376 | |||
BC144722.1 | - | 3 | 435 |