Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135100.2 | NP_001128572.1 | 2, 3 | 607 | ||
NM_029646.3 | NP_083922.1 | 2, 3 | 607 | |||
GenBank mRNA | AK008082.1 | - | 2, 3 | 555 | ||
BC016254.1 | - | 2, 3 | 586 | 4912097 | ||
EU636995.1 | - | 1, 2 | 24 | |||
EU636996.1 | - | 1, 2 | 24 |