Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190156.1 | NP_001177085.1 | 5 | 730 | ||
NM_029655.3 | NP_083931.2 | 5 | 730 | |||
GenBank mRNA | AK011015.1 | - | 5 | 705 | ||
AK038971.1 | - | 5 | 731 | |||
AK052487.1 | - | 5 | 687 | |||
AK170966.1 | - | 5 | 677 | |||
BC087942.1 | - | 5 | 588 | 30303462 |