Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081054.2 | NP_001074523.1 | 4 | 432 | ||
GenBank mRNA | AK012351.1 | - | 4 | 433 | ||
AK038801.1 | - | 4 | 441 | |||
AK041347.1 | - | 4 | 437 | |||
AK170225.1 | - | 4 | 377 | |||
BC070459.1 | - | 4 | 423 | 30605375 |