Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054043.3 | NP_473384.1 | 9 | 688 | ||
GenBank mRNA | AK049637.1 | - | 9 | 688 | ||
AK049688.1 | - | 9 | 688 | |||
AK051269.1 | - | 9 | 538 | |||
AK136156.1 | - | 5 | 496 | |||
AK153928.1 | - | 9 | 561 | |||
AK169846.1 | - | 9 | 561 | |||
BC111809.1 | - | 10 | 621 | 40045350 |