Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081114.1 | NP_001074583.1 | 4 | 516 | ||
GenBank mRNA | BC138413.1 | - | 3 | 435 | 8861433 |