Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029706.1 | NP_083982.1 | 6, 7 | 653 | ||
GenBank mRNA | AK003061.1 | - | 6, 7 | 592 | ||
AK008678.1 | - | 3, 4 | 325 | |||
BC138759.1 | - | 6, 7 | 611 | 8861781 | ||
BC138762.1 | - | 6, 7 | 611 | 8861784 |