Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198415.2 | NP_940807.1 | 5 | 708 | ||
GenBank mRNA | AK009042.1 | - | 3 | 412 | ||
BC061221.1 | - | 5 | 708 | 30296538 |