Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133766.3 | NP_598527.2 | 15 | 1910 | ||
GenBank mRNA | AB158474.1 | - | 15 | 1914 | ||
AK018695.1 | - | 10 | 1125 | |||
AK030074.1 | - | 15 | 1911 | |||
AK032461.1 | - | 15 | 1883 | |||
AK044000.1 | - | 15 | 1904 | |||
AK044475.1 | - | 15 | 1906 | |||
AK129065.1 | - | 15 | 1888 | |||
AK161250.1 | - | 15 | 1895 |