Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082484.2 | NP_001075953.1 | 9 | 1474 | ||
NM_029721.2 | NP_083997.1 | 9 | 1474 | |||
GenBank mRNA | AK017836.1 | - | 6 | 545 | ||
AK039051.1 | - | 2 | 145 | |||
AK041201.1 | - | 2 | 178 | |||
AK122296.1 | - | 9 | 1286 | |||
AK147452.1 | - | 9 | 1461 | |||
BC053495.1 | - | 9 | 1286 |