Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290523.1 | NP_001277452.1 | 5, 6 | 640 | ||
NM_001290524.1 | NP_001277453.1 | 5, 6 | 640 | |||
NM_029730.4 | NP_084006.2 | 5, 6 | 640 | |||
GenBank mRNA | AK010480.1 | - | 5, 6 | 562 | ||
AK031405.1 | - | 5, 6 | 565 | |||
AK035985.1 | - | 5, 6 | 600 | |||
AK135188.1 | - | 5, 6 | 595 | |||
AK156572.1 | - | 5, 6 | 583 | |||
BC026425.1 | - | 5, 6 | 566 | 4223175 |