Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081082.2 | NP_001074551.1 | 5 | 622 | ||
GenBank mRNA | BC119800.1 | - | 5 | 550 |