Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290759.1 | NP_001277688.1 | 22 | 3745 | ||
NM_001290760.1 | NP_001277689.1 | 22 | 3748 | |||
NM_029745.2 | NP_084021.2 | 21 | 3697 | |||
GenBank mRNA | AK012224.1 | - | 1 | 567 | ||
AK031992.1 | - | 7 | 1167 | |||
AK035457.1 | - | 21 | 3662 | |||
AK088860.1 | - | 16 | 2647 | |||
AK172999.1 | - | 23 | 3685 | |||
AK183366.1 | - | 1 | 68 | |||
AK190655.1 | - | 1 | 68 | |||
AK191880.1 | - | 1 | 68 | |||
AK197752.1 | - | 1 | 68 | |||
AK212412.1 | - | 1 | 68 | |||
AK214088.1 | - | 1 | 68 | |||
AK219911.1 | - | 1 | 58 | |||
BC048085.1 | - | 12 | 2011 | |||
BC058596.1 | - | 9 | 1282 | |||
BC062928.1 | - | 21 | 3684 | 6854669 | ||
BC065080.1 | - | 21 | 3676 | 6826791 | ||
BC065130.1 | - | 21 | 3640 |