Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB095735.1 | - | 11 | 1130 | ||
AB095736.1 | - | 11 | 1130 | |||
AK054412.1 | - | 11 | 1120 | |||
AK133177.1 | - | 11 | 1129 | |||
AK154594.1 | - | 12 | 1170 | |||
AK161401.1 | - | 8 | 731 | |||
AK164249.1 | - | 11 | 1130 | |||
AK164999.1 | - | 11 | 1129 | |||
AK166351.1 | - | 11 | 1138 | |||
AK169601.1 | - | 11 | 1129 | |||
BC060208.1 | - | 11 | 1089 |