Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB095735.1 | - | 14 | 1752 | ||
AB095736.1 | - | 14 | 1752 | |||
AK054412.1 | - | 14 | 1742 | |||
AK133177.1 | - | 14 | 1751 | |||
AK154594.1 | - | 15 | 1793 | |||
AK161401.1 | - | 11 | 1353 | |||
AK164249.1 | - | 14 | 1752 | |||
AK166351.1 | - | 14 | 1760 | |||
AK169601.1 | - | 14 | 1777 | |||
BC060208.1 | - | 14 | 1711 |