Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153177.3 | NP_694817.2 | 15 | 2313 | ||
GenBank mRNA | AB081474.1 | - | 15 | 2058 | ||
AK030018.1 | - | 15 | 2329 | |||
AK032475.1 | - | 4 | 638 | |||
AK129395.2 | - | 11 | 1495 | |||
BC096023.1 | - | 15 | 2095 | 30607807 |