Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002894.2 | NP_001002894.2 | 3 | 940 | ||
GenBank mRNA | AY596199.1 | - | 2 | 816 | ||
AY673647.1 | - | 3 | 940 | |||
BC141199.1 | - | 3 | 924 | 9055811 |