Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029775.2 | NP_084051.1 | 6, 7 | 784 | ||
GenBank mRNA | AK013933.1 | - | 6, 7 | 802 | ||
AK014737.1 | - | 6, 7 | 826 | |||
AK134862.1 | - | 831 | ||||
BC020048.1 | - | 6, 7 | 820 | |||
BC020089.1 | - | 6, 7 | 784 | 4013755 | ||
BC049641.1 | - | 973 |