Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252459.1 | NP_001239388.1 | 26 | 3407 | ||
NM_001252460.1 | NP_001239389.1 | 26 | 3462 | |||
NM_133769.3 | NP_598530.2 | 26 | 3170 | |||
GenBank mRNA | AB093288.2 | - | 25 | 3042 | ||
AF334144.1 | - | 26 | 3105 | |||
AK030397.1 | - | 26 | 3107 | |||
AK147224.1 | - | 26 | 3408 | |||
AK147586.1 | - | 26 | 3163 | |||
AK147632.1 | - | 26 | 3104 | |||
AK147971.1 | - | 26 | 3111 | |||
BC038080.1 | - | 9 | 982 | |||
BC049232.1 | - | 1 | 51 |