Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039179.2 | NP_001034268.1 | 6, 7 | 2371 | ||
NM_001039180.2 | NP_001034269.1 | 7, 8 | 2267 | |||
NM_029791.4 | NP_084067.1 | 6, 7 | 2371 | |||
GenBank mRNA | AJ250106.1 | - | 6, 7 | 2330 | ||
AK003456.1 | - | 1, 2 | 95 | |||
AK122348.1 | - | 6, 7 | 2344 | |||
AK156407.1 | - | 6, 7 | 2326 | |||
AK171300.1 | - | 7, 8 | 2225 | |||
BC032198.1 | - | 6, 7 | 2328 | 5324113 |