Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039179.2 | NP_001034268.1 | 5 | 1783 | ||
NM_001039180.2 | NP_001034269.1 | 6 | 1679 | |||
NM_029791.4 | NP_084067.1 | 5 | 1783 | |||
GenBank mRNA | AJ250106.1 | - | 5 | 1742 | ||
AK122348.1 | - | 5 | 1756 | |||
AK156407.1 | - | 5 | 1738 | |||
AK171300.1 | - | 6 | 1637 | |||
AK189233.1 | - | 1 | 240 | |||
BC032198.1 | - | 5 | 1740 | 5324113 |