Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290649.1 | NP_001277578.1 | 8 | 909 | ||
NM_029793.2 | NP_084069.1 | 9 | 984 | |||
GenBank mRNA | AK002954.1 | - | 9 | 928 | ||
AK008972.1 | - | 9 | 883 | |||
AK028516.1 | - | 9 | 904 | |||
AK044987.1 | - | 9 | 932 | |||
BC024637.1 | - | 9 | 906 | |||
BC037037.1 | - | 5 | 421 | |||
BC043090.1 | - | 9 | 849 | 6418353 | ||
BC053068.1 | - | 8 | 837 | 6405595 | ||
BC080733.1 | - | 8 | 909 | 30110235 |