Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029802.4 | NP_084078.3 | 8 | 1160 | ||
GenBank mRNA | AK009100.1 | - | 8 | 1098 | ||
AK154772.1 | - | 8 | 1126 | |||
BC013794.1 | - | 8 | 1071 | 2648132 | ||
BC022942.1 | - | 8 | 1084 | 3583149 | ||
BC028310.1 | - | 8 | 1075 | |||
BC034520.1 | - | 8 | 1141 | 3964827 | ||
BC079603.1 | - | 5 | 2333 |