Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256115.1 | NP_001243044.1 | 16 | 1853 | ||
NM_001256116.1 | NP_001243045.1 | 15 | 1709 | |||
NM_001256117.1 | NP_001243046.1 | 14 | 1595 | |||
NM_001256118.1 | NP_001243047.1 | 12 | 1433 | |||
NM_024451.2 | NP_077771.1 | 17 | 1964 | |||
GenBank mRNA | AF343752.1 | - | 1 | 69 | ||
AK014585.1 | - | 14 | 1593 | |||
AK036187.1 | - | 16 | 1830 | |||
AK160281.1 | - | 17 | 1922 | |||
AK167686.1 | - | 17 | 1936 | |||
AK203786.1 | - | 4 | 366 | |||
AY682989.1 | - | 15 | 1689 | |||
BC047928.1 | - | 17 | 1931 | 5321879 | ||
BC048156.1 | - | 17 | 1933 | 5698402 | ||
HQ402597.1 | - | 12 | 1413 |