Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198163.1 | NP_937806.1 | 6 | 661 | ||
GenBank mRNA | AB232631.1 | - | 6 | 501 | ||
AK156576.1 | - | 6 | 652 | |||
AK185540.1 | - | 2 | 40 | |||
AK191987.1 | - | 2 | 40 | |||
AK195760.1 | - | 2 | 40 | |||
AK197903.1 | - | 2 | 40 | |||
AK200288.1 | - | 40 | ||||
AK201126.1 | - | 2 | 40 | |||
AK207070.1 | - | 2 | 40 | |||
BC030941.1 | - | 4 | 352 | |||
BC056466.1 | - | 6 | 661 | 5717246 |