Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163787.1 | NP_001157259.1 | 10 | 1529 | ||
NM_029939.3 | NP_084215.2 | 10 | 1526 | |||
GenBank mRNA | AK031175.1 | - | 10 | 1527 | ||
BC051179.1 | - | 10 | 1446 | |||
BC057069.1 | - | 10 | 1432 | 5690884 |