Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163787.1 | NP_001157259.1 | 3 | 559 | ||
NM_029939.3 | NP_084215.2 | 3 | 559 | |||
GenBank mRNA | AK021166.1 | - | 3 | 560 | ||
AK031175.1 | - | 3 | 560 | |||
BC051179.1 | - | 3 | 478 | |||
BC057069.1 | - | 3 | 462 | 5690884 |