Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029961.2 | NP_084237.1 | 15, 16 | 1950 | ||
GenBank mRNA | AK139832.1 | - | 3, 4 | 428 | ||
AY766239.1 | - | 15, 16 | 1914 | |||
JQ655148.1 | - | 2, 3 | 1429 |