Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020737.1 | - | 4 | 429 | ||
AK030269.1 | - | 6 | 880 | |||
AK041560.1 | - | 5 | 568 | |||
BC125478.1 | - | 6 | 865 | 40129993 | ||
BC125480.1 | - | 6 | 865 | 40129995 |