Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081222.1 | NP_001074691.1 | 8 | 3018 | ||
GenBank mRNA | AK037793.1 | - | 1 | 1212 | ||
AK040576.1 | - | 8 | 846 | |||
AK049589.1 | - | 7 | 734 | |||
AK139549.1 | - | 8 | 2742 | |||
AK145333.1 | - | 8 | 3019 | |||
AK145870.1 | - | 7 | 666 | |||
AK173297.1 | - | 6 | 2398 | |||
BC008220.1 | - | 7 | 890 | |||
BC076613.1 | - | 8 | 2748 | |||
BC151069.1 | - | 8 | 2974 | 9087982 | ||
BC151077.1 | - | 8 | 2974 | 9087990 | ||
BC172153.1 | - | 9 | 3096 |