Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029985.2 | NP_084261.1 | 7 | 1258 | ||
GenBank mRNA | AK020847.1 | - | 7 | 1064 | ||
AK169814.1 | - | 5 | 716 | |||
BC027203.1 | - | 7 | 1190 | 3710091 | ||
BC031894.1 | - | 7 | 1162 | 5362699 |