Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175268.4 | NP_780477.2 | 5 | 1459 | ||
NM_212473.1 | NP_997638.1 | 5 | 1522 | |||
GenBank mRNA | AK031576.1 | - | 4 | 1303 | ||
AK051299.1 | - | 5 | 1523 | |||
AK147350.1 | - | 5 | 1855 | |||
AK147725.1 | - | 5 | 1651 | |||
AK165022.1 | - | 737 | ||||
BC035523.1 | - | 5 | 1454 | 5400742 |