Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081030.2 | NP_001074499.2 | 39 | 5683 | ||
NM_001170561.1 | NP_001164032.1 | 40 | 5761 | |||
GenBank mRNA | AK076039.1 | - | 7 | 928 | ||
AK129485.1 | - | 39 | 5627 | |||
AK147332.1 | - | 39 | 5640 | |||
BC029156.1 | - | 10 | 1319 | |||
BC098209.1 | - | 20 | 3026 | |||
BC157935.1 | - | 39 | 5573 | 9088115 | ||
BC158013.1 | - | 40 | 5651 | 9007320 | ||
BC172094.1 | - | 39 | 5573 | 9054380 |