Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252470.1 | NP_001239399.1 | 3 | 518 | ||
NM_153679.2 | NP_710146.1 | 3 | 520 | |||
GenBank mRNA | AF320000.1 | - | 3 | 443 | ||
AK035790.1 | - | 3 | 484 | |||
AK087747.1 | - | 3 | 392 | |||
BC066155.1 | - | 3 | 442 | 30089333 | ||
BC083317.1 | - | 3 | 401 |