Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030138.2 | NP_084414.1 | 22 | 2432 | ||
GenBank mRNA | AB490692.1 | - | 23 | 2206 | ||
AK029343.1 | - | 22 | 2356 | |||
AK129041.1 | - | 22 | 2409 | |||
AK147319.1 | - | 21 | 2304 | |||
BC046455.1 | - | 9 | 975 |